2-(Methylamino)-5-nitrobenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR300182-1G
APOSOR300182-1GEA
101
EUR
APOSOR300182-1G
2-(Methylamino)-5-nitrobenzoic acid
2-(Methylamino)-5-nitrobenzoic acid
Formel:
C₈H₈N₂O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00956206 CAS-Nummer: 3484-33-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...