2-Amino-5-bromoquinazolin-4-ol

Lieferant: Apollo Scientific
Achtung

OR300183-250MG
APOSOR300183-250MGEA 126 EUR
APOSOR300183-250MG
2-Amino-5-bromoquinazolin-4-ol
2-Amino-5-bromoquinazolin-4-ol
Formel: C₈H₆BrN₃O
Molecular Weight: 240,057 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18379108
CAS-Nummer: 937668-63-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR