2-Amino-7-bromoquinazolin-4-ol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-7-bromoquinazolin-4(3H)-one

OR300185-250MG
APOSOR300185-250MGEA 231 EUR
APOSOR300185-250MG
2-Amino-7-bromoquinazolin-4-ol
2-Amino-7-bromoquinazolin-4-ol
Formel: C₈H₆BrN₃O
Molecular Weight: 240,057 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18379110
CAS-Nummer: 885277-56-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR