4-(Methylamino)bicyclo[2.2.2]octane-1-carbonitrile

Lieferant: Apollo Scientific
Gefahr

OR300190-250MG
APOSOR300190-250MGEA 190 EUR
APOSOR300190-250MG
4-(Methylamino)bicyclo[2.2.2]octane-1-carbonitrile
4-(Methylamino)bicyclo[2.2.2]octane-1-carbonitrile
Formel: C₁₀H₁₆N₂
Molecular Weight: 164,247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31536721
CAS-Nummer: 2187435-31-8
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR