4-(Methylamino)bicyclo[2.2.2]octane-1-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR300191-250MG
APOSOR300191-250MGEA
377
EUR
APOSOR300191-250MG
4-(Methylamino)bicyclo[2.2.2]octane-1-carboxylic acid
4-(Methylamino)bicyclo[2.2.2]octane-1-carboxylic acid
Formel:
C₁₀H₁₇NO₂ Molecular Weight: 183,247 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31536722 CAS-Nummer: 792136-91-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...