4-Bromo-7-methoxy-1-methyl-1H-pyrrolo[2,3-c]pyridine

Lieferant: Apollo Scientific
Achtung

OR300193-250MG
APOSOR300193-250MGEA 97.3 EUR
APOSOR300193-250MG
4-Bromo-7-methoxy-1-methyl-1H-pyrrolo[2,3-c]pyridine
4-Bromo-7-methoxy-1-methyl-1H-pyrrolo[2,3-c]pyridine
Formel: C₉H₉BrN₂O
Molecular Weight: 241,085 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31536724
CAS-Nummer: 2103352-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR