5-Bromo-3-methyl-1H-indole-7-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR300196-250MG
APOSOR300196-250MGEA
246
EUR
APOSOR300196-250MG
5-Bromo-3-methyl-1H-indole-7-carbonitrile
5-Bromo-3-methyl-1H-indole-7-carbonitrile
Formel:
C₁₀H₇BrN₂ Molecular Weight: 235,08 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD31536726 CAS-Nummer: 1221448-64-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...