7-Methoxy-1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-c]pyridine

Lieferant: Apollo Scientific
Achtung

OR300200-250MG
APOSOR300200-250MGEA 295 EUR
APOSOR300200-250MG
7-Methoxy-1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-c]pyridine
7-Methoxy-1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-c]pyridine
Formel: C₁₅H₂₁BN₂O₃
Molecular Weight: 288,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30294840
CAS-Nummer: 1627713-58-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR