7-tert-Butyl-5-bromo-1H-indole

Lieferant: Apollo Scientific
Achtung

OR300201-250MG
APOSOR300201-250MGEA 377 EUR
APOSOR300201-250MG
7-tert-Butyl-5-bromo-1H-indole
7-tert-Butyl-5-bromo-1H-indole
Formel: C₁₂H₁₄BrN
Molecular Weight: 252,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28658735
CAS-Nummer: 1219832-07-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR