Benzyl 4-(aminomethyl)bicyclo[2.2.2]octan-1-ylcarbamate

Lieferant: Apollo Scientific
Achtung

OR300202-250MG
APOSOR300202-250MGEA 580 EUR
APOSOR300202-250MG
Benzyl 4-(aminomethyl)bicyclo[2.2.2]octan-1-ylcarbamate
Benzyl 4-(aminomethyl)bicyclo[2.2.2]octan-1-ylcarbamate
Formel: C₁₇H₂₄N₂O₂
Molecular Weight: 288,385 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29042274
CAS-Nummer: 2227272-82-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR