Benzyl 4-(aminomethyl)bicyclo[2.2.2]octan-1-ylmethylcarbamate

Lieferant: Apollo Scientific
Achtung

OR300203-250MG
APOSOR300203-250MGEA 387 EUR
APOSOR300203-250MG
Benzyl 4-(aminomethyl)bicyclo[2.2.2]octan-1-ylmethylcarbamate
Benzyl 4-(aminomethyl)bicyclo[2.2.2]octan-1-ylmethylcarbamate
Formel: C₁₈H₂₆N₂O₂
Molecular Weight: 302.411 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31536729
CAS-Nummer: 2187435-32-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR