Benzyl 4-cyanobicyclo[2.2.2]octan-1-ylcarbamate

Lieferant: Apollo Scientific
Achtung

OR300204-250MG
APOSOR300204-250MGEA 536 EUR
APOSOR300204-250MG
Benzyl 4-cyanobicyclo[2.2.2]octan-1-ylcarbamate
Benzyl 4-cyanobicyclo[2.2.2]octan-1-ylcarbamate
Formel: C₁₇H₂₀N₂O₂
Molecular Weight: 284,353 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31536730
CAS-Nummer: 1252672-88-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR