Benzyl 4-cyanobicyclo[2.2.2]octan-1-ylmethylcarbamate

Lieferant: Apollo Scientific
Achtung

OR300205-250MG
APOSOR300205-250MGEA 376 EUR
APOSOR300205-250MG
Benzyl 4-cyanobicyclo[2.2.2]octan-1-ylmethylcarbamate
Benzyl 4-cyanobicyclo[2.2.2]octan-1-ylmethylcarbamate
Formel: C₁₈H₂₂N₂O₂
Molecular Weight: 298,379 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31536731
CAS-Nummer: 2187434-96-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR