Bicyclo[2.2.2]octane-1-carboxylic acid 95+%

Lieferant: Apollo Scientific
Achtung

OR300206-50MG
APOSOR300206-50MGEA 21.9 EUR
APOSOR300206-50MG
Bicyclo[2.2.2]octane-1-carboxylic acid 95+%
Bicyclo[2.2.2]octane-1-carboxylic acid
Formel: C₉H₁₄O₂
Molecular Weight: 154,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00218755
CAS-Nummer: 699-55-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR