Methyl 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-6-carboxylate

Lieferant: Apollo Scientific
Achtung

OR300207-250MG
APOSOR300207-250MGEA 572 EUR
APOSOR300207-250MG
Methyl 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-6-carboxylate
Methyl 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-6-carboxylate
Formel: C₁₇H₂₂BNO₄
Molecular Weight: 315.172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31536732
CAS-Nummer: 2103352-55-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR