Methyl 5-chloro-2-methyl-1H-indole-3-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR300209-250MG
APOSOR300209-250MGEA
190
EUR
APOSOR300209-250MG
Methyl 5-chloro-2-methyl-1H-indole-3-carboxylate
Methyl 5-chloro-2-methyl-1H-indole-3-carboxylate
Formel:
C₁₁H₁₀ClNO₂ Molecular Weight: 223,656 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23159660 CAS-Nummer: 762288-06-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...