N,N-Dimethyl-1H-pyrrolo[2,3-c]pyridin-7-amine

Lieferant: Apollo Scientific
Gefahr

OR300210-250MG
APOSOR300210-250MGEA 633 EUR
APOSOR300210-250MG
N,N-Dimethyl-1H-pyrrolo[2,3-c]pyridin-7-amine
N,N-Dimethyl-1H-pyrrolo[2,3-c]pyridin-7-amine
Formel: C₉H₁₁N₃
Molecular Weight: 161,204 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31536734
CAS-Nummer: 2103352-79-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR