N,N-Dimethyl-1H-thieno[2,3-c]pyrazol-3-amine

Lieferant: Apollo Scientific
Achtung

OR300211-250MG
APOSOR300211-250MGEA 265 EUR
APOSOR300211-250MG
N,N-Dimethyl-1H-thieno[2,3-c]pyrazol-3-amine
N,N-Dimethyl-1H-thieno[2,3-c]pyrazol-3-amine
Formel: C₇H₉N₃S
Molecular Weight: 167,231 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31536735
CAS-Nummer: 2227272-46-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR