6-Ethynyl-8-phenylimidazo[1,2-a]pyrazine

Lieferant: Apollo Scientific
Achtung

OR300214-100MG
APOSOR300214-100MGEA 223 EUR
APOSOR300214-100MG
6-Ethynyl-8-phenylimidazo[1,2-a]pyrazine
6-Ethynyl-8-phenylimidazo[1,2-a]pyrazine
Formel: C₁₄H₉N₃
Lagertemperatur: Kühlschrank
MDL: MFCD31561247
CAS-Nummer: 2227272-60-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR