4-Bromo-7-methoxy-1H-pyrrolo[2,3-c]pyridine 98%

Lieferant: Apollo Scientific

OR300216-250MG
APOSOR300216-250MGEA 116 EUR
APOSOR300216-250MG
4-Bromo-7-methoxy-1H-pyrrolo[2,3-c]pyridine 98%
4-Bromo-7-methoxy-1H-pyrrolo[2,3-c]pyridine
Formel: C₈H₇BrN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD11845544
CAS-Nummer: 425380-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR