5-Bromo-2-(tetrahydro-2H-pyran-4-yl)isoindolin-1-one
Lieferant: Apollo Scientific
|
Achtung
|
OR300218-250MG
APOSOR300218-250MGEA
190
EUR
APOSOR300218-250MG
5-Bromo-2-(tetrahydro-2H-pyran-4-yl)isoindolin-1-one
5-Bromo-2-(tetrahydro-2H-pyran-4-yl)isoindolin-1-one
Formel:
C₁₃H₁₄BrNO₂ Molecular Weight: 296,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31561251 CAS-Nummer: 1614234-06-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...