6-Iodo-8-phenylimidazo[1,2-a]pyrazine
Lieferant: Apollo Scientific
|
Achtung
|
OR300224-250MG
APOSOR300224-250MGEA
451
EUR
APOSOR300224-250MG
6-Iodo-8-phenylimidazo[1,2-a]pyrazine
6-Iodo-8-phenylimidazo[1,2-a]pyrazine
Formel:
C₁₂H₈IN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD31561255 CAS-Nummer: 2227272-86-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...