1-Bromo-3-(2,2-dibromovinyl)-5-methylbenzene
Lieferant: Apollo Scientific
|
Gefahr
|
OR300226-250MG
APOSOR300226-250MGEA
301
EUR
APOSOR300226-250MG
1-Bromo-3-(2,2-dibromovinyl)-5-methylbenzene
1-Bromo-3-(2,2-dibromovinyl)-5-methylbenzene
Formel:
C₉H₇Br₃ Molecular Weight: 354,864 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31561257 CAS-Nummer: 2227272-74-2 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...