1-Bromo-3-(2-bromoethynyl)-5-isopropylbenzene

Lieferant: Apollo Scientific
Achtung

OR300227-250MG
APOSOR300227-250MGEA 358 EUR
APOSOR300227-250MG
1-Bromo-3-(2-bromoethynyl)-5-isopropylbenzene
1-Bromo-3-(2-bromoethynyl)-5-isopropylbenzene
Formel: C₁₁H₁₀Br₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31561259
CAS-Nummer: 2227272-72-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR