1-Bromo-3-(2-bromoethynyl)-5-methylbenzene
Lieferant: Apollo Scientific
|
Gefahr
|
OR300228-250MG
APOSOR300228-250MGEA
339
EUR
APOSOR300228-250MG
1-Bromo-3-(2-bromoethynyl)-5-methylbenzene
1-Bromo-3-(2-bromoethynyl)-5-methylbenzene
Formel:
C₉H₆Br₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD31561261 CAS-Nummer: 2227272-64-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...