1-(3-Bromo-5-isopropylphenyl)ethanone

Lieferant: Apollo Scientific
Achtung

OR300229-250MG
APOSOR300229-250MGEA 301 EUR
APOSOR300229-250MG
1-(3-Bromo-5-isopropylphenyl)ethanone
1-(3-Bromo-5-isopropylphenyl)ethanone
Formel: C₁₁H₁₃BrO
Molecular Weight: 241,124 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31561263
CAS-Nummer: 1782470-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR