1-(3-Bromo-5-isopropylphenyl)ethanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3-Bromo-5-isopropylphenyl)ethan-1-amine hydrochloride

OR300231-250MG
APOSOR300231-250MGEA 358 EUR
APOSOR300231-250MG
1-(3-Bromo-5-isopropylphenyl)ethanamine hydrochloride
1-(3-Bromo-5-isopropylphenyl)ethanamine hydrochloride
Formel: C₁₁H₁₆BrN·ClH
Molecular Weight: 278,616 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31561265
CAS-Nummer: 2227272-85-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR