3-Bromo-5-isopropylbenzonitrile

Lieferant: Apollo Scientific
Achtung

OR300233-250MG
APOSOR300233-250MGEA 415 EUR
APOSOR300233-250MG
3-Bromo-5-isopropylbenzonitrile
3-Bromo-5-isopropylbenzonitrile
Formel: C₁₀H₁₀BrN
Molecular Weight: 224,097 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29094527
CAS-Nummer: 1369854-24-9
UN: 3082
ADR: 9+EHS,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR