(3-Bromo-5-isopropylphenyl)methanamine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR300234-250MG
APOSOR300234-250MGEA
487
EUR
APOSOR300234-250MG
(3-Bromo-5-isopropylphenyl)methanamine hydrochloride
(3-Bromo-5-isopropylphenyl)methanamine hydrochloride
Formel:
C₁₀H₁₄BrN·ClH Molecular Weight: 264,59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31561267 CAS-Nummer: 2227272-65-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...