tert-Butyl 3-bromo-5-isopropylbenzylcarbamate

Lieferant: Apollo Scientific
Achtung

OR300235-250MG
APOSOR300235-250MGEA 567 EUR
APOSOR300235-250MG
tert-Butyl 3-bromo-5-isopropylbenzylcarbamate
tert-Butyl 3-bromo-5-isopropylbenzylcarbamate
Formel: C₁₅H₂₂BrNO₂
Molecular Weight: 328,245 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31561268
CAS-Nummer: 2227272-55-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR