8-Chloro-6-iodoimidazo[1,2-a]pyrazine

Lieferant: Apollo Scientific
Achtung

OR300240-1G OR300240-250MG
APOSOR300240-1GEA 277 EUR
APOSOR300240-1G APOSOR300240-250MG
8-Chloro-6-iodoimidazo[1,2-a]pyrazine
8-Chloro-6-iodoimidazo[1,2-a]pyrazine
Formel: C₆H₃ClIN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD31561275
CAS-Nummer: 2227272-49-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR