5-Bromo-3-phenylpyrazin-2-amine

Lieferant: Apollo Scientific
Achtung

OR300242-250MG
APOSOR300242-250MGEA 68.3 EUR
APOSOR300242-250MG
5-Bromo-3-phenylpyrazin-2-amine
5-Bromo-3-phenylpyrazin-2-amine
Formel: C₁₀H₈BrN₃
Molecular Weight: 250,095 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12547094
CAS-Nummer: 344940-70-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR