5-Ethynyl-3-phenylpyrazin-2-amine

Lieferant: Apollo Scientific
Achtung

OR300244-1G
APOSOR300244-1GEA 289 EUR
APOSOR300244-1G
5-Ethynyl-3-phenylpyrazin-2-amine
5-Ethynyl-3-phenylpyrazin-2-amine
Formel: C₁₂H₉N₃
Lagertemperatur: Kühlschrank
MDL: MFCD31561280
CAS-Nummer: 2227272-78-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR