2-Bromo-1-(3-bromo-5-methylphenyl)ethanone

Lieferant: Apollo Scientific
Gefahr

OR300245-1G
APOSOR300245-1GEA 877 EUR
APOSOR300245-1G
2-Bromo-1-(3-bromo-5-methylphenyl)ethanone
2-Bromo-1-(3-bromo-5-methylphenyl)ethanone
Formel: C₉H₈Br₂O
Molecular Weight: 291,967 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23711607
CAS-Nummer: 260430-27-1
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR