tert-Butyl 3-bromo-5-methylbenzylcarbamate

Lieferant: Apollo Scientific
Achtung

OR300247-1G OR300247-250MG
APOSOR300247-1GEA 618 EUR
APOSOR300247-1G APOSOR300247-250MG
tert-Butyl 3-bromo-5-methylbenzylcarbamate
tert-Butyl 3-bromo-5-methylbenzylcarbamate
Formel: C₁₃H₁₈BrNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD30531038
CAS-Nummer: 1998726-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR