2-Methoxyquinoline-6-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR300253-250MG
APOSOR300253-250MGEA
246
EUR
APOSOR300253-250MG
2-Methoxyquinoline-6-carbonitrile
2-Methoxyquinoline-6-carbonitrile
Formel:
C₁₁H₈N₂O Molecular Weight: 184,194 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27921733 CAS-Nummer: 99471-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...