6-Bromo-2-ethoxyquinoline

Lieferant: Apollo Scientific
Achtung

OR300254-250MG
APOSOR300254-250MGEA 214 EUR
APOSOR300254-250MG
6-Bromo-2-ethoxyquinoline
6-Bromo-2-ethoxyquinoline
Formel: C₁₁H₁₀BrNO
Molecular Weight: 252,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27944300
CAS-Nummer: 1363386-54-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR