Methyl 4,5,6,7-tetrahydrothiazolo[4,5-c]pyridine-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR300258-250MG
APOSOR300258-250MGEA 719 EUR
APOSOR300258-250MG
Methyl 4,5,6,7-tetrahydrothiazolo[4,5-c]pyridine-2-carboxylate
Methyl 4,5,6,7-tetrahydrothiazolo[4,5-c]pyridine-2-carboxylate
Formel: C₈H₁₀N₂O₂S
Molecular Weight: 198.242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19703765
CAS-Nummer: 1256806-74-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR