tert-Butyl 2-amino-6,7-dihydrothiazolo[4,5-c]pyridine-5(4H)-carboxylate

Lieferant: Apollo Scientific
Achtung

OR300259-250MG
APOSOR300259-250MGEA 246 EUR
APOSOR300259-250MG
tert-Butyl 2-amino-6,7-dihydrothiazolo[4,5-c]pyridine-5(4H)-carboxylate
tert-Butyl 2-amino-6,7-dihydrothiazolo[4,5-c]pyridine-5(4H)-carboxylate
Formel: C₁₁H₁₇N₃O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD11045435
CAS-Nummer: 1002355-91-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR