2-(Hydroxymethyl)cyclohexa-2,5-diene-1,4-dione
Lieferant: Apollo Scientific
|
Achtung
|
OR300264-250MG
APOSOR300264-250MGEA
226
EUR
APOSOR300264-250MG
2-(Hydroxymethyl)cyclohexa-2,5-diene-1,4-dione
2-(Hydroxymethyl)cyclohexa-2,5-diene-1,4-dione
Formel:
C₇H₆O₃ Schmelzpunkt: 76…77 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01663152 CAS-Nummer: 644-17-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...