7-Bromo-2-mercaptoquinazolin-4(3H)-one 95+%

Lieferant: Apollo Scientific
Achtung

OR300268-250MG
APOSOR300268-250MGEA 183 EUR
APOSOR300268-250MG
7-Bromo-2-mercaptoquinazolin-4(3H)-one 95+%
7-Bromo-2-mercaptoquinazolin-4(3H)-one
Formel: C₈H₅BrN₂OS
Molecular Weight: 257,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31561260
CAS-Nummer: 1463501-47-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR