5-Bromo-2-mercaptoquinazolin-4(3H)-one

Lieferant: Apollo Scientific
Achtung

OR300269-250MG
APOSOR300269-250MGEA 238 EUR
APOSOR300269-250MG
5-Bromo-2-mercaptoquinazolin-4(3H)-one
5-Bromo-2-mercaptoquinazolin-4(3H)-one
Formel: C₈H₅BrN₂OS
Molecular Weight: 257,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31561262
CAS-Nummer: 2231674-69-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR