tert-Butyl 5-formylindoline-1-carboxylate

Lieferant: Apollo Scientific
Achtung

OR300270-1G OR300270-250MG
APOSOR300270-1GEA 290 EUR
APOSOR300270-1G APOSOR300270-250MG
tert-Butyl 5-formylindoline-1-carboxylate
tert-Butyl 5-formylindoline-1-carboxylate
Formel: C₁₄H₁₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD12408236
CAS-Nummer: 879887-32-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR