2-Aminobenzo[d]thiazole-5-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR300283-1G OR300283-250MG
APOSOR300283-1GEA 314 EUR
APOSOR300283-1G APOSOR300283-250MG
2-Aminobenzo[d]thiazole-5-carbonitrile
2-Aminobenzo[d]thiazole-5-carbonitrile
Formel: C₈H₅N₃S
Molecular Weight: 175.21 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 105314-08-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR