2-Chloro-4-methylquinoline-3-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR30030-1G
APOSOR30030-1GEA 139 EUR
APOSOR30030-1G
2-Chloro-4-methylquinoline-3-carbonitrile
2-Chloro-4-methylquinoline-3-carbonitrile
Formel: C₁₁H₇ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00833327
CAS-Nummer: 101617-94-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR