4-Methyl-2-phenylthiazol-5-ylboronic acid, pinacol ester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Methyl-2-phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole
OR300303-250MG
APOSOR300303-250MGEA
197
EUR
APOSOR300303-250MG
4-Methyl-2-phenylthiazol-5-ylboronic acid, pinacol ester
4-Methyl-2-phenylthiazol-5-ylboronic acid, pinacol ester
Formel:
C₁₆H₂₀BNO₂S Molecular Weight: 301,211 g/mol Schmelzpunkt: 75…76 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05865106 CAS-Nummer: 690632-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...