4-Bromo-1H-indole-6-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR300306-250MG
APOSOR300306-250MGEA 270 EUR
APOSOR300306-250MG
4-Bromo-1H-indole-6-carbonitrile
4-Bromo-1H-indole-6-carbonitrile
Formel: C₉H₅BrN₂
Molecular Weight: 221,053 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095209
CAS-Nummer: 374633-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR