1-Chloro-4-iodo-2,7-naphthyridine

Lieferant: Apollo Scientific
Achtung

OR300339-250MG
APOSOR300339-250MGEA 358 EUR
APOSOR300339-250MG
1-Chloro-4-iodo-2,7-naphthyridine
1-Chloro-4-iodo-2,7-naphthyridine
Formel: C₈H₄ClIN₂
Molecular Weight: 290,488 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17015945
CAS-Nummer: 1234616-02-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR