1,6-Naphthyridine-5-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR300345-1G OR300345-250MG
APOSOR300345-1GEA 1080 EUR
APOSOR300345-1G APOSOR300345-250MG
1,6-Naphthyridine-5-carbonitrile
1,6-Naphthyridine-5-carbonitrile
Formel: C₉H₅N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06762717
CAS-Nummer: 28694-40-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR