3-Bromo-5-chloroisonicotinonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR300370-1G
OR300370-250MG
APOSOR300370-1GEA
404
EUR
APOSOR300370-1G
APOSOR300370-250MG
3-Bromo-5-chloroisonicotinonitrile
3-Bromo-5-chloroisonicotinonitrile
Formel:
C₆H₂BrClN₂ Molecular Weight: 217.45 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22380416 CAS-Nummer: 1335052-66-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...